APO455827168 Apollo Scientific Ltd

2,6-Dimethylpiperidine

  • MDL number MFCD00005986
  • Cas Number : 504-03-0
  • Molecular Weight : 113.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point <-20
Synonym 2,6-Lupetidine
Density 0.832
Flash Point 11
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 126-129
Product Link https://store.apolloscientific.co.uk/product/26-dimethylpiperidine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethylpiperidine
pictograms GHS02;GHS05
hstatements H225;H314
Availability details for
Quantity
  • Availability
  • Add to Cart