APO455827169 Apollo Scientific Ltd

2-Acetyl-6-methylpyridine

  • MDL number MFCD06658293
  • Cas Number : 6940-57-4
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(6-Methylpyridin-2-yl)ethan-1-one, 6-Acetyl-2-picoline
Flash Point 84
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 209
Product Link https://store.apolloscientific.co.uk/product/2-acetyl-6-methylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-acetyl-6-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart