APO455827174 Apollo Scientific Ltd

2-(Methoxymethyl)pyridine

  • MDL number MFCD00030825
  • Cas Number : 23579-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methoxymethylpyridine

Safety Information

signalword Danger
pcodes P310;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxymethylpyridine
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart