APO455827175 Apollo Scientific Ltd

3-Chloro-2-hydroxypyridine

  • MDL number MFCD01646109
  • Cas Number : 13466-35-8
  • Molecular Weight : 129.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177
Synonym 3-Chloropyridin-2-ol
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-hydroxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-hydroxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart