APO455827177 Apollo Scientific Ltd

3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulphonyl chloride

  • MDL number MFCD06797486
  • Cas Number : 10185-62-3
  • Molecular Weight : 258.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 67-69
Iupac Name 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride
Inchi Code 1S/C9H7ClN2O3S/c1-6-11-9(12-15-6)7-3-2-4-8(5-7)16(10,13)14/h2-5H,1H3
Inchi Key YPMVPGZXUILSPT-UHFFFAOYSA-N

Description

Synonym 3-[3-(Chlorosulphonyl)phenyl]-5-methyl-1,2,4-oxadiazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 67-69
Product Link https://store.apolloscientific.co.uk/product/3-5-methyl-124-oxadiazol-3-ylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-5-methyl-124-oxadiazol-3-ylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart