APO455827179 Apollo Scientific Ltd

4-Propylpyridine

  • MDL number MFCD00014639
  • Cas Number : 1122-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Pyridin-4-yl)propane
Density 0.93
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 190
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42743

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42743
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart