APO455827189 Apollo Scientific Ltd

2-Phenylacrylic acid

  • MDL number MFCD00046531
  • Cas Number : 492-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107
Synonym 2-Phenylprop-2-enoic acid, Atropic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 267
Product Link https://store.apolloscientific.co.uk/product/2-phenylacrylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-phenylacrylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart