APO455827195 Apollo Scientific Ltd

3-Amino-2,6-dibromo-5-ethoxypyridine

  • MDL number MFCD09864810
  • Cas Number : 1000018-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Dibromo-5-ethoxypyridin-3-amine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38737

Safety Information

signalword Warning
pcodes P260;P280;P301+P312;P302+P352;P304+P340;P361+P364
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38737
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart