APO455827196 Apollo Scientific Ltd

Methyl 3-iodo-4-methylbenzoate

  • MDL number MFCD00230583
  • Cas Number : 90347-66-3
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26-29
Synonym 2-Iodo-4-(methoxycarbonyl)toluene, Methyl 3-iodo-p-toluate
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-iodo-4-methylbenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-iodo-4-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart