APO455827209 Apollo Scientific Ltd

3,5-Dibromo-2-methylthiophene

  • MDL number MFCD07781197
  • Cas Number : 29421-73-6
  • Molecular Weight : 255.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dibromo-2-methylthiophene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dibromo-2-methylthiophene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart