APO455827225 Apollo Scientific Ltd

4-(3-Aminobenzyl)piperazine, N1-BOC protected

  • MDL number MFCD06658979
  • Cas Number : 361345-40-6
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Synonym tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-3-aminobenzylpiperazine-n1-boc-protected

Safety Information

signalword Danger
pcodes P260;P280;P308+P313
MSDS https://store.apolloscientific.co.uk/product/4-3-aminobenzylpiperazine-n1-boc-protected
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart