APO455827227 Apollo Scientific Ltd

Methyl 5-bromo-6-chloronicotinate

  • MDL number MFCD03844855
  • Cas Number : 78686-77-8
  • Molecular Weight : 250.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-79
Synonym Methyl 5-bromo-6-chloropyridine-3-carboxylate, 3-Bromo-2-chloro-5-(methoxycarbonyl)pyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-bromo-6-chloronicotinate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-bromo-6-chloronicotinate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart