APO455827229 Apollo Scientific Ltd

2-Bromo-6-hydrazinopyridine

  • MDL number MFCD09743754
  • Cas Number : 26944-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-118
Synonym (6-Bromopyridin-2-yl)hydrazine
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-hydrazinopyridine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-hydrazinopyridine
pictograms GHS05;GHS06
hstatements H301;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart