APO455827239 Apollo Scientific Ltd

Thiomorpholine 1,1-dioxide

  • MDL number MFCD05861623
  • Cas Number : 39093-93-1
  • Molecular Weight : 135.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-71
Synonym 1,4-Thiazinane 1,1-dioxide, 1,1-Dioxidothiomorpholine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 155/1mm
Product Link https://store.apolloscientific.co.uk/product/thiomorpholine-11-dioxide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/thiomorpholine-11-dioxide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart