APO455827252 Apollo Scientific Ltd

6-Bromobenzo[b]thiophene-2-carboxylic acid

  • MDL number MFCD07371540
  • Cas Number : 19075-58-2
  • Molecular Weight : 257.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262-265
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromobenzobthiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromobenzobthiophene-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart