APO455827256 Apollo Scientific Ltd

3-(1,3-Oxazol-5-yl)benzoic acid

  • MDL number MFCD06797481
  • Cas Number : 252928-82-8
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258-260
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-13-oxazol-5-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-13-oxazol-5-ylbenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart