APO455827257 Apollo Scientific Ltd

4-(Piperazin-1-yl)piperidine, N-BOC protected

  • MDL number MFCD04973999
  • Cas Number : 177276-41-4
  • Molecular Weight : 269.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 4-(piperazin-1-yl)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(piperazin-1-yl)piperidine
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-piperazin-1-ylpiperidine-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-piperazin-1-ylpiperidine-n-boc-protected
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart