APO455827259 Apollo Scientific Ltd

6-Chlorobenzo[b]thiophene-2-carboxylic acid

  • MDL number MFCD07377059
  • Cas Number : 26018-73-5
  • Molecular Weight : 212.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 271-274
Synonym 2-Carboxy-6-chlorobenzo[b]thiophene, 2-Carboxy-6-chloro-1-benzothiophene, 2-Carboxy-6-chlorothianaphthene
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chlorobenzobthiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chlorobenzobthiophene-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart