APO455827263 Apollo Scientific Ltd

Ethyl 5-bromo-1-methyl-1H-indole-2-carboxylate

  • MDL number MFCD09027165
  • Cas Number : 91844-20-1
  • Molecular Weight : 282.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Synonym 5-Bromo-2-(ethoxycarbonyl)-1-methyl-1H-indole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-bromo-1-methyl-1h-indole-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-bromo-1-methyl-1h-indole-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart