APO455827265 Apollo Scientific Ltd

2-Phenyl-1,3-thiazolidine

  • MDL number MFCD00128238
  • Cas Number : 4569-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Synonym 2-Phenyl-1,3-thiazolane, (1,3-Thiazolidin-2-yl)benzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-phenyl-13-thiazolidine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-phenyl-13-thiazolidine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart