APO455827271 Apollo Scientific Ltd

5-Bromo-6-phenylpyridazin-3(2H)-one

  • MDL number MFCD02083298
  • Cas Number : 90766-97-5
  • Molecular Weight : 251.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234-237
Synonym 5-Bromo-2,3-dihydro-3-oxo-6-phenylpyridazine, 5-Bromo-2,3-dihydro-3-oxo-6-phenyl-1,2-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-6-phenylpyridazin-32h-one

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-6-phenylpyridazin-32h-one
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart