APO455827278 Apollo Scientific Ltd

1-(Ethoxycarbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD07339354
  • Cas Number : 118133-15-6
  • Molecular Weight : 201.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Synonym 4-Carboxy-1-(ethoxycarbonyl)piperidine, Ethyl 4-carboxypiperidine-1-carboxylate
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-ethoxycarbonylpiperidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-ethoxycarbonylpiperidine-4-carboxylic-acid
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart