APO455827280 Apollo Scientific Ltd

2-Amino-4,5-dimethyl-1,3-thiazole

  • MDL number MFCD00462425
  • Cas Number : 2289-75-0
  • Molecular Weight : 128.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-69
Synonym 4,5-Dimethyl-1,3-thiazol-2-amine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-45-dimethyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-45-dimethyl-13-thiazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart