APO455827285 Apollo Scientific Ltd

2-(4-Methylbenzoyl)benzoic acid

  • MDL number MFCD00020287
  • Cas Number : 85-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-138
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38695

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38695
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart