APO455827286 Apollo Scientific Ltd

Methyl 3-aminoisonicotinate

  • MDL number MFCD02082535
  • Cas Number : 55279-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-aminopyridine-4-carboxylate, 3-Amino-4-(methoxycarbonyl)pyridine
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38698

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=38698
Availability details for
Quantity
  • Availability
  • Add to Cart