APO455827293 Apollo Scientific Ltd

2,6-Dichloro-3-formylbenzeneboronic acid

  • MDL number MFCD06796322
  • Cas Number : 1218790-87-4
  • Molecular Weight : 218.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-154
Synonym 3-Borono-2,4-dichlorobenzaldehyde
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-3-formylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-3-formylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart