APO455827294 Apollo Scientific Ltd

3-(Isopropoxycarbonyl)benzeneboronic acid

  • MDL number MFCD02093047
  • Cas Number : 342002-80-6
  • Molecular Weight : 208.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 153-154
Iupac Name 3-(isopropoxycarbonyl)phenylboronic acid
Inchi Code 1S/C10H13BO4/c1-7(2)15-10(12)8-4-3-5-9(6-8)11(13)14/h3-7,13-14H,1-2H3
Inchi Key HXVODQWMURGQMP-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 153-154
Product Link https://store.apolloscientific.co.uk/product/3-isopropoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/3-isopropoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart