APO455827297 Apollo Scientific Ltd

Pyridine-4-boronic acid hydrochloride

  • MDL number MFCD06201888
  • Cas Number : 913835-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28586

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28586
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart