APO455827302 Apollo Scientific Ltd

5-Amino-1-(4-methylphenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00052944
  • Cas Number : 103646-82-8
  • Molecular Weight : 198.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-120
Synonym 4-(5-Amino-4-cyano-1H-pyrazol-1-yl)toluene, 5-Amino-4-cyano-1-(4-methylphenyl)-1H-pyrazole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-4-methylphenyl-1h-pyrazole-4-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-4-methylphenyl-1h-pyrazole-4-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart