APO455827307 Apollo Scientific Ltd

5-(Chloromethyl)-2-(4-methoxyphenyl)pyridine

  • MDL number MFCD06200955
  • Cas Number : 886361-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-49
Synonym 4-[5-(Chloromethyl)pyridin-2-yl]anisole, 6-(4-Methoxyphenyl)nicotinyl chloride
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28683

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28683
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart