APO455827313 Apollo Scientific Ltd

2-Hydrazino-1H-benzimidazole

  • MDL number MFCD00513603
  • Cas Number : 15108-18-6
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 224-225
Iupac Name 2-hydrazino-1H-benzimidazole
Inchi Code 1S/C7H8N4/c8-11-7-9-5-3-1-2-4-6(5)10-7/h1-4H,8H2,(H2,9,10,11)
Inchi Key PXUYWNZBZXDJEU-UHFFFAOYSA-N

Description

Synonym (1H-Benzimidazol-2-yl)hydrazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 224-225
Product Link https://store.apolloscientific.co.uk/product/2-hydrazino-1h-benzimidazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydrazino-1h-benzimidazole
pictograms GHS07
hstatements H302;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart