APO455827317 Apollo Scientific Ltd

6-(4-Methoxyphenyl)pyridin-3-amine

  • MDL number MFCD05864840
  • Cas Number : 52057-98-4
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Synonym 3-Amino-6-(4-methoxyphenyl)pyridine, 4-(5-Aminopyridin-2-yl)anisole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-4-methoxyphenylpyridin-3-amine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-4-methoxyphenylpyridin-3-amine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart