APO455827319 Apollo Scientific Ltd

Methyl imidazo[1,2-a]pyridine-6-carboxylate

  • MDL number MFCD06200984
  • Cas Number : 136117-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-148
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-imidazo12-apyridine-6-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-imidazo12-apyridine-6-carboxylate
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart