APO455827326 Apollo Scientific Ltd

tert-Butyl-3-(4-nitrophenoxy)tetrahydro-1(2H)-pyridinecarboxylate

  • MDL number MFCD05865146
  • Cas Number : 690632-16-7
  • Molecular Weight : 322.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(4-nitrophenoxy)-1-piperidinecarboxylate
Inchi Code 1S/C16H22N2O5/c1-16(2,3)23-15(19)17-10-4-5-14(11-17)22-13-8-6-12(7-9-13)18(20)21/h6-9,14H,4-5,10-11H2,1-3H3
Inchi Key XPOFZYJUWZHERT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-3-4-nitrophenoxytetrahydro-12h-pyridinecarboxylate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-3-4-nitrophenoxytetrahydro-12h-pyridinecarboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart