APO455827339 Apollo Scientific Ltd

2-(Chloroacetyl)-3,4-dibromothiophene

  • MDL number MFCD06411631
  • Cas Number : 62673-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-98
Synonym 2-Chloro-1-(3,4-dibromothien-2-yl)ethan-1-one, 2-(2-Chloroethanoyl)-3,4-dibromothiophene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28732

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28732
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart