APO455827342 Apollo Scientific Ltd

(5-Formyl-2-methoxyphenoxy)(phenyl)acetic acid

  • MDL number MFCD06411628
  • Cas Number : 885949-43-9
  • Molecular Weight : 286.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-161
Synonym 2-(5-Formyl-2-methoxyphenoxy)-2-phenylethanoic acid, 3-[Carboxy(phenyl)methoxy]-4-methoxybenzaldehyde
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-formyl-2-methoxyphenoxyphenylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-formyl-2-methoxyphenoxyphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart