APO455827343 Apollo Scientific Ltd

Hydroxy(2-hydroxynaphth-1-yl)acetic acid

  • MDL number MFCD06411618
  • Cas Number : 624722-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Synonym 2-Hydroxy-2-(2-hydroxynaphth-1-yl)ethanoic acid, 1-[Carboxy(hydroxy)methyl]-2-hydroxynaphthalene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28748

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28748
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart