APO455827344 Apollo Scientific Ltd

2-Amino-7-cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD07366548
  • Cas Number : 885949-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Cyclopropyl[1,2,4]triazolo[1,5-a]pyrimidin-2-amine, 1-(2-Amino[1,2,4]triazolo[1,5-a]pyrimidin-7-yl)cyclopropane
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28750

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=28750
Availability details for
Quantity
  • Availability
  • Add to Cart