APO455827347 Apollo Scientific Ltd

1-(Methylsulphonyl)piperidine-4-carboxylic acid

  • MDL number MFCD03821886
  • Cas Number : 280772-00-1
  • Molecular Weight : 207.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Synonym 4-Carboxy-1-(methylsulphonyl)piperidine, 4-Carboxypiperidin-1-yl methyl sulphone
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-methylsulphonylpiperidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylsulphonylpiperidine-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart