APO455827357 Apollo Scientific Ltd

5-Methylbenzo[b]thiophene-2-boronic acid

  • MDL number MFCD06200866
  • Cas Number : 136099-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100
Synonym 2-Borono-5-methylbenzo[b]thiophene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28739

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28739
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart