APO455827359 Apollo Scientific Ltd

5-(Aminomethyl)-1-methyl-1H-imidazole

  • MDL number MFCD06200858
  • Cas Number : 486414-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1-Methyl-1H-imidazol-5-yl)methylamine
Purity 97
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 100-104/0.3mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28741

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28741
pictograms GHS05;GHS07
hstatements H302;H312;H314
Availability details for
Quantity
  • Availability
  • Add to Cart