APO455827363 Apollo Scientific Ltd

1-Aminoadamantane

  • MDL number MFCD00074732
  • Cas Number : 768-94-5
  • Molecular Weight : 151.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-208
Synonym Adamantan-1-amine, Tricyclo[3.3.1.1~3,7~]decan-1-amine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-aminoadamantane_OR14310

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-aminoadamantane_OR14310
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart