APO455827365 Apollo Scientific Ltd

4-[4-(Hydroxymethyl)phenyl]pyridine

  • MDL number MFCD04114576
  • Cas Number : 217192-22-8
  • Molecular Weight : 185.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 172-176
Iupac Name [4-(4-pyridinyl)phenyl]methanol
Inchi Code 1S/C12H11NO/c14-9-10-1-3-11(4-2-10)12-5-7-13-8-6-12/h1-8,14H,9H2
Inchi Key JTQRRRUVWBLIFL-UHFFFAOYSA-N

Description

Synonym (4-Pyridin-4-ylphenyl)methanol
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 172-176
Product Link https://store.apolloscientific.co.uk/product/4-4-hydroxymethylphenylpyridine

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-hydroxymethylphenylpyridine
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart