APO455827384 Apollo Scientific Ltd

Ethyl 3-aminothieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD00173981
  • Cas Number : 52505-46-1
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-185
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-aminothieno23-bpyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-aminothieno23-bpyridine-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart