APO455827391 Apollo Scientific Ltd

2-Amino-4-(4-bromophenyl)pyrimidine-5-carbonitrile

  • MDL number MFCD06659571
  • Cas Number : 749885-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258-260
Synonym 2-Amino-4-(4-bromophenyl)-5-cyanopyrimidine, 2-Amino-4-(4-bromophenyl)-5-cyano-1,3-diazine, 1-(2-Amino-5-cyanopyrimidin-4-yl)-4-bromobenzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28827

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28827
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart