APO455827393 Apollo Scientific Ltd

(4-Aminophenyl)[4-(pyrimidin-2-yl)piperazin-1-yl]methanone

  • MDL number MFCD06659558
  • Cas Number : 885949-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216
Synonym 1-(4-Aminobenzoyl)-4-(pyrimidin-2-yl)piperazine, 4-{[4-(Pyrimidin-2-yl)piperazin-1-yl]carbonyl}aniline, 2-[4-(4-Aminobenzoyl)piperazin-1-yl]pyrimidine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28830

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28830
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart