APO455827400 Apollo Scientific Ltd

Methyl 6-amino-5-bromonicotinate

  • MDL number MFCD06659561
  • Cas Number : 180340-70-9
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-169
Synonym Methyl 6-amino-5-bromopyridine-3-carboxylate, Methyl 2-amino-3-bromopyridine-5-carboxylate, 2-Amino-3-bromo-5-(methoxycarbonyl)pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-6-amino-5-bromonicotinate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-6-amino-5-bromonicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart