APO455827426 Apollo Scientific Ltd

5-Bromo-1H-indazole-3-carboxylic acid

  • MDL number MFCD05663979
  • Cas Number : 1077-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290-292
Synonym 5-Bromo-3-carboxy-1H-indazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-1h-indazole-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-1h-indazole-3-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart