APO455827428 Apollo Scientific Ltd

Pyridine-2-amidoxime

  • MDL number MFCD00085159
  • Cas Number : 1772-01-6
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 120-124
Iupac Name N'-hydroxy-2-pyridinecarboximidamide
Inchi Code 1S/C6H7N3O/c7-6(9-10)5-3-1-2-4-8-5/h1-4,6H,7H2
Inchi Key IAUNAOJNUMBOJS-UHFFFAOYSA-N

Description

Synonym N'-Hydroxypyridine-2-carboximidamide, N'-Hydroxypyridine-2-carboxamidine, Pyridine-2-carboxamide oxime
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 120-124
Product Link https://store.apolloscientific.co.uk/product/pyridine-2-amidoxime

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/pyridine-2-amidoxime
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart