APO455827434 Apollo Scientific Ltd

Methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate

  • MDL number MFCD06660133
  • Cas Number : 208465-72-9
  • Molecular Weight : 175.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 72-76
Iupac Name methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate
Inchi Code 1S/C6H6ClNO3/c1-10-6(9)4-3-11-5(2-7)8-4/h3H,2H2,1H3
Inchi Key CMUKPCIZFMTLKD-UHFFFAOYSA-N

Description

Synonym 2-(Chloromethyl)-4-(methoxycarbonyl)-1,3-oxazole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 72-76
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloromethyl-13-oxazole-4-carboxylate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloromethyl-13-oxazole-4-carboxylate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart